Product Details

SNP ID
rs11178937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:71766185 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTGATTTACCTGGGCATATTGTA[A/G]ATGTGCTTATCTATAATAACAGTAT
Phenotype
MIM: 612398
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB21 PubMed Links

Gene Details

Gene
RAB21
Gene Name
RAB21, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014999.3 Intron NP_055814.1

View Full Product Details