Product Details

SNP ID
rs11634420
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:91102298 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCGTGCACTACAAGAAGCATGAG[C/T]GTGGATGCTCAAACAGACCTAGGTG
Phenotype
MIM: 185861
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SV2B PubMed Links

Gene Details

Gene
SV2B
Gene Name
synaptic vesicle glycoprotein 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167580.1 482 Intron NP_001161052.1
NM_001323031.1 482 Intron NP_001309960.1
NM_001323032.1 482 Intron NP_001309961.1
NM_001323033.1 482 Intron NP_001309962.1
NM_001323034.1 482 Intron NP_001309963.1
NM_001323036.1 482 Intron NP_001309965.1
NM_001323037.1 482 Intron NP_001309966.1
NM_001323038.1 482 Intron NP_001309967.1
NM_001323039.1 482 UTR 5 NP_001309968.1
NM_001323040.1 482 UTR 5 NP_001309969.1
NM_014848.5 482 Intron NP_055663.1
XM_005254998.3 482 Intron XP_005255055.1
XM_017022761.1 482 Intron XP_016878250.1
XM_017022762.1 482 Intron XP_016878251.1

View Full Product Details