Product Details

SNP ID
rs10920056
Assay Type
Functionally tested
NCBI dbSNP Submissions
27
Location
Chr.1:200873505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCACTGCGCACCCCGCGCTGCGC[G/T]CTGGCCTCGTGCTGCGGCGTCTGGG
Phenotype
MIM: 602174
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR25 PubMed Links

Gene Details

Gene
GPR25
Gene Name
G protein-coupled receptor 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005298.3 551 Silent Mutation GCG,GCT A156A NP_005289.2

View Full Product Details