Product Details

SNP ID
rs10775505
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:24163527 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATTTCCCACCAGGATCTAGCAAG[C/T]TACTTGACAGAAGCAGGGTCTTCTA
Phenotype
MIM: 612135 MIM: 606730
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CABYR PubMed Links

Gene Details

Gene
CABYR
Gene Name
calcium binding tyrosine phosphorylation regulated
There are no transcripts associated with this gene.

Gene
OSBPL1A
Gene Name
oxysterol binding protein like 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242508.1 Intron NP_001229437.1
NM_018030.4 Intron NP_060500.3
NM_080597.3 Intron NP_542164.2
XM_006722380.3 Intron XP_006722443.1
XM_006722382.3 Intron XP_006722445.1
XM_017025530.1 Intron XP_016881019.1
XM_017025531.1 Intron XP_016881020.1
XM_017025532.1 Intron XP_016881021.1
XM_017025533.1 Intron XP_016881022.1
XM_017025534.1 Intron XP_016881023.1

View Full Product Details