Product Details

SNP ID
rs12327412
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:23992873 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTTGCTACAACAGAGGCAGGATT[C/G]TGAATTTATTTCCAGCTGCCCATCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TTC39C PubMed Links

Gene Details

Gene
TTC39C
Gene Name
tetratricopeptide repeat domain 39C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135993.1 101 Intron NP_001129465.1
NM_001243425.1 101 Intron NP_001230354.1
NM_001292030.1 101 Intron NP_001278959.1
NM_153211.3 101 UTR 5 NP_694943.2
XM_005258198.4 101 Intron XP_005258255.1
XM_006722392.3 101 Intron XP_006722455.1
XM_011525814.2 101 Intron XP_011524116.1
XM_017025550.1 101 UTR 5 XP_016881039.1
Gene
TTC39C-AS1
Gene Name
TTC39C antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details