Product Details

SNP ID
rs12459655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57908290 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGTGTGGTGTACTCGTAATCTCA[G/T]CTCCTTGGGAGGCTGAGGTAGGAGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF417 PubMed Links

Gene Details

Gene
ZNF417
Gene Name
zinc finger protein 417
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297734.1 2780 Intron NP_001284663.1
NM_152475.2 2780 Intron NP_689688.2
XM_011526473.2 2780 UTR 3 XP_011524775.1
XM_017026315.1 2780 Intron XP_016881804.1

View Full Product Details