Product Details

SNP ID
rs11802972
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:42692628 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCCAGTTCATTGTAACCTTTGT[C/T]CCCTTTGTCATTAAACGCTAGAGCT
Phenotype
MIM: 154030
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
YBX1 PubMed Links

Gene Details

Gene
YBX1
Gene Name
Y-box binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004559.4 Intron NP_004550.2

View Full Product Details