Product Details

SNP ID
rs12479154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:120346414 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCGAGTGGACGGCGACTTCCTG[C/G]AGGCGGTGAAGCGGCACATCTTGAG
Phenotype
MIM: 147390
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
INHBB PubMed Links

Gene Details

Gene
INHBB
Gene Name
inhibin beta B subunit
There are no transcripts associated with this gene.

Gene
LOC105373989
Gene Name
putative cuticle collagen 91
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017005445.1 Intron XP_016860934.1

View Full Product Details