Product Details

SNP ID
rs11110200
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:100030800 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCTGTCTAAATGATTTCAGAATT[A/T]AAAAAAAACCCAGATTACTATTTTT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
UHRF1BP1L PubMed Links

Gene Details

Gene
UHRF1BP1L
Gene Name
UHRF1 binding protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006947.1 Intron NP_001006948.1
NM_015054.1 Intron NP_055869.1
XM_005268738.2 Intron XP_005268795.1
XM_005268739.3 Intron XP_005268796.1
XM_011538054.2 Intron XP_011536356.1
XM_017019047.1 Intron XP_016874536.1
XM_017019048.1 Intron XP_016874537.1
XM_017019049.1 Intron XP_016874538.1
XM_017019050.1 Intron XP_016874539.1
XM_017019051.1 Intron XP_016874540.1

View Full Product Details