Product Details

SNP ID
rs11722858
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:133966875 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAGCTTTTGAAATGGTTTCAAAA[A/G]CATATTAGCAATGCAGACCAGTTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PABPC4L PubMed Links

Gene Details

Gene
PABPC4L
Gene Name
poly(A) binding protein cytoplasmic 4 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001114734.1 Intron NP_001108206.2
XM_017007727.1 Intron XP_016863216.1

View Full Product Details