Product Details

SNP ID
hCV30868815
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:81991237 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCCCCCGCCCACACACCTCAGCT[C/T]TACGTAATACATACTCACATGACAA
Phenotype
MIM: 616164
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SDR42E1 PubMed Links

Gene Details

Gene
SDR42E1
Gene Name
short chain dehydrogenase/reductase family 42E, member 1
There are no transcripts associated with this gene.

View Full Product Details