Product Details

SNP ID
rs4254322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:81779948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACACTCGCGATGCCCGCTCCGCA[A/G]CTGTCTGCGTAATTATGCGTGTGTC
Phenotype
MIM: 600220
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105369213 PubMed Links

Gene Details

Gene
LOC105369213
Gene Name
uncharacterized LOC105369213
There are no transcripts associated with this gene.

Gene
PLCG2
Gene Name
phospholipase C gamma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002661.4 Intron NP_002652.2

View Full Product Details