Product Details

SNP ID
rs12127737
Assay Type
Functionally tested
NCBI dbSNP Submissions
38
Location
Chr.1:43936893 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCCTCAAAGCTGAGAGGCCCCT[A/G]CCGGTGGGTGATGGATATCATCCCC
Phenotype
MIM: 603886 MIM: 610411 MIM: 606494
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARTN PubMed Links

Gene Details

Gene
ARTN
Gene Name
artemin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136215.1 1573 UTR 3 NP_001129687.1
NM_057090.2 1573 UTR 3 NP_476431.2
NM_057091.2 1573 UTR 3 NP_476432.2
Gene
IPO13
Gene Name
importin 13
There are no transcripts associated with this gene.

Gene
ST3GAL3
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 3
There are no transcripts associated with this gene.

View Full Product Details