Product Details

SNP ID
rs11573601
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:44821439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGCCTTGGAGGTTGCTCTGGAA[C/T]CTGGCTCCTTTCCCTGGGTCCACAA
Phenotype
MIM: 603673
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTBD19 PubMed Links

Gene Details

Gene
BTBD19
Gene Name
BTB domain containing 19
There are no transcripts associated with this gene.

Gene
PTCH2
Gene Name
patched 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166292.1 Intron NP_001159764.1
NM_003738.4 Intron NP_003729.3

View Full Product Details