Product Details

SNP ID
rs12731245
Assay Type
Functionally tested
NCBI dbSNP Submissions
35
Location
Chr.1:45021579 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGACTGTTATATCTTATAAAGGA[C/G]AAATACATTAAGTAAAGATAAAACA
Phenotype
MIM: 613521
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
UROD PubMed Links

Gene Details

Gene
UROD
Gene Name
uroporphyrinogen decarboxylase
There are no transcripts associated with this gene.

Gene
ZSWIM5
Gene Name
zinc finger SWIM-type containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020883.1 Intron NP_065934.1
XM_011541861.2 Intron XP_011540163.1
XM_017001913.1 Intron XP_016857402.1

View Full Product Details