Product Details

SNP ID
rs12049503
Assay Type
Functionally tested
NCBI dbSNP Submissions
50
Location
Chr.1:45555884 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGGACATGGCGAATCTCAGATGC[A/C]TATACATACTAGTGGAGAGGTCCAG
Phenotype
MIM: 103830
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AKR1A1 PubMed Links

Gene Details

Gene
AKR1A1
Gene Name
aldo-keto reductase family 1 member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202413.1 Intron NP_001189342.1
NM_001202414.1 Intron NP_001189343.1
NM_006066.3 Intron NP_006057.1
NM_153326.2 Intron NP_697021.1
XM_011540491.2 Intron XP_011538793.1
XM_011540492.2 Intron XP_011538794.1

View Full Product Details