Product Details

SNP ID
rs12037459
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:45629287 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGACCAAGAGCTCAGACCATAGCTC[G/T]GGAACACAGCCTTGTGGTTCTGATA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC17 PubMed Links

Gene Details

Gene
CCDC17
Gene Name
coiled-coil domain containing 17
There are no transcripts associated with this gene.

Gene
GPBP1L1
Gene Name
GC-rich promoter binding protein 1 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021639.4 Intron NP_067652.1
XM_017002019.1 Intron XP_016857508.1

View Full Product Details