Product Details

SNP ID
rs11563905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:90346737 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCAAGAAGGTTTCCTGGCCTGTTG[C/T]AGCCATGGTGCATTGCAGTTGCGTG
Phenotype
MIM: 610920
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTPBP10 PubMed Links

Gene Details

Gene
GTPBP10
Gene Name
GTP binding protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042717.2 73 UTR 5 NP_001036182.1
NM_033107.3 73 UTR 5 NP_149098.2
Gene
LOC101927446
Gene Name
uncharacterized LOC101927446
There are no transcripts associated with this gene.

View Full Product Details