Product Details

SNP ID
rs13158027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:34844284 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGCCTCAAGAAATGTTGCTTCCC[C/T]ATCTACACAACCTGGTTTGCCCACG
Phenotype
MIM: 616344
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTC23L PubMed Links

Gene Details

Gene
TTC23L
Gene Name
tetratricopeptide repeat domain 23 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317949.1 Intron NP_001304878.1
NM_144725.3 Intron NP_653326.3
XM_011513984.2 Intron XP_011512286.1
XM_011513985.2 Intron XP_011512287.1
XM_011513987.2 Intron XP_011512289.1
XM_017009119.1 Intron XP_016864608.1
XM_017009120.1 Intron XP_016864609.1
XM_017009121.1 Intron XP_016864610.1
XM_017009122.1 Intron XP_016864611.1
XM_017009123.1 Intron XP_016864612.1
XM_017009124.1 Intron XP_016864613.1
XM_017009125.1 Intron XP_016864614.1
XM_017009126.1 Intron XP_016864615.1

View Full Product Details