Product Details

SNP ID
rs12054220
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:136958739 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCCTGCCAGTGAGATTACATA[A/G]TTTTCTCTGCTCCACTCCTGGTTGC
Phenotype
MIM: 605621 MIM: 600508
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL20RB PubMed Links

Gene Details

Gene
IL20RB
Gene Name
interleukin 20 receptor subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144717.3 Intron NP_653318.2
XM_006713665.3 Intron XP_006713728.1
XM_011512910.2 Intron XP_011511212.1
XM_011512911.2 Intron XP_011511213.1
XM_017006632.1 Intron XP_016862121.1
Gene
NCK1
Gene Name
NCK adaptor protein 1
There are no transcripts associated with this gene.

View Full Product Details