Product Details
- SNP ID
-
hCV30883253
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:114446533 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGGTCTTCCTCTTTCTCTTTCCTT[C/T]TTACAAGACCTGCCTGATTTATTTT
- Phenotype
-
MIM: 608209
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DPP10
PubMed Links
Gene Details
- Gene
- DPP10
- Gene Name
- dipeptidyl peptidase like 10
There are no transcripts associated with this gene.
View Full Product Details