Product Details

SNP ID
rs12354500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73441652 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATAGTGCTATCTCAAACTGTGTTC[C/T]TCAGTACTATGCAAGATAAATGACC
Phenotype
MIM: 614773 MIM: 114106
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MSS51 PubMed Links

Gene Details

Gene
MSS51
Gene Name
MSS51 mitochondrial translational activator
There are no transcripts associated with this gene.

Gene
PPP3CB
Gene Name
protein phosphatase 3 catalytic subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142353.2 Intron NP_001135825.1
NM_001142354.2 Intron NP_001135826.1
NM_001289968.1 Intron NP_001276897.1
NM_001289969.1 Intron NP_001276898.1
NM_021132.3 Intron NP_066955.1
XM_005269944.1 Intron XP_005270001.1
XM_017016379.1 Intron XP_016871868.1

View Full Product Details