Product Details

SNP ID
rs11000569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:73265356 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAAGAAGAAGAAGAAGAGATTC[A/G]TGTATATTTTCTTCTTTTTCCTTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CFAP70 PubMed Links

Gene Details

Gene
CFAP70
Gene Name
cilia and flagella associated protein 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145170.3 Intron NP_660153.3
XM_005269489.2 Intron XP_005269546.1
XM_006717604.2 Intron XP_006717667.1
XM_006717605.2 Intron XP_006717668.1
XM_006717609.2 Intron XP_006717672.1
XM_006717610.2 Intron XP_006717673.1
XM_006717611.3 Intron XP_006717674.1
XM_011539212.2 Intron XP_011537514.1
XM_017015620.1 Intron XP_016871109.1
XM_017015621.1 Intron XP_016871110.1
XM_017015622.1 Intron XP_016871111.1
XM_017015623.1 Intron XP_016871112.1
XM_017015624.1 Intron XP_016871113.1
XM_017015625.1 Intron XP_016871114.1
XM_017015626.1 Intron XP_016871115.1
XM_017015627.1 Intron XP_016871116.1
XM_017015628.1 Intron XP_016871117.1
XM_017015629.1 Intron XP_016871118.1
XM_017015630.1 Intron XP_016871119.1
XM_017015631.1 Intron XP_016871120.1
XM_017015632.1 Intron XP_016871121.1
XM_017015633.1 Intron XP_016871122.1
Gene
DNAJC9-AS1
Gene Name
DNAJC9 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details