Product Details

SNP ID
rs12039961
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:203307418 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGGAGCAGCCCCTCCAAGAACTAC[A/G]TGATGGCAGTCTCCAGCTAGGCCCT
Phenotype
MIM: 601597
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTG2 PubMed Links

Gene Details

Gene
BTG2
Gene Name
BTG anti-proliferation factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006763.2 528 Missense Mutation ATG,GTG M153V NP_006754.1
Gene
LINC01136
Gene Name
long intergenic non-protein coding RNA 1136
There are no transcripts associated with this gene.

View Full Product Details