Product Details

SNP ID
rs11582753
Assay Type
Functionally tested
NCBI dbSNP Submissions
51
Location
Chr.1:203014166 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGCATAAAGCTGGAATTCGGAGTC[A/G]GAAACTCTTGGTTCTATTTCCCTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100506747 PubMed Links

Gene Details

Gene
LOC100506747
Gene Name
alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase-like protein LOC641515 homolog
There are no transcripts associated with this gene.

Gene
TMEM183A
Gene Name
transmembrane protein 183A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322955.1 Intron NP_001309884.1
NM_001322956.1 Intron NP_001309885.1
NM_001322957.1 Intron NP_001309886.1
NM_001322958.1 Intron NP_001309887.1
NM_001322959.1 Intron NP_001309888.1
NM_138391.4 Intron NP_612400.3

View Full Product Details