Product Details
- SNP ID
-
rs4757757
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:19122597 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGTTTGTGGAAGCTTTCTCTGCCC[C/T]TAGGTTACCTTGTAAACAGAGTATA
- Phenotype
-
MIM: 612815
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZDHHC13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141990021] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZDHHC13
- Gene Name
- zinc finger DHHC-type containing 13
View Full Product Details