Product Details

SNP ID
rs11600760
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18534480 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGTTACAAAAATACGTGATCTCA[G/T]AAAATGCATAAAATATGCACATTTA
Phenotype
MIM: 601387 MIM: 610985
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TSG101 PubMed Links

Gene Details

Gene
TSG101
Gene Name
tumor susceptibility 101
There are no transcripts associated with this gene.

Gene
UEVLD
Gene Name
UEV and lactate/malate dehyrogenase domains
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040697.2 Intron NP_001035787.1
NM_001261382.1 Intron NP_001248311.1
NM_001261383.1 Intron NP_001248312.1
NM_001261384.1 Intron NP_001248313.1
NM_001261385.1 Intron NP_001248314.1
NM_001261386.1 Intron NP_001248315.1
NM_001297771.1 Intron NP_001284700.1
NM_018314.4 Intron NP_060784.3
XM_006718255.2 Intron XP_006718318.1
XM_011520201.2 Intron XP_011518503.1
XM_017017985.1 Intron XP_016873474.1
XM_017017986.1 Intron XP_016873475.1

View Full Product Details