Product Details

SNP ID
rs13377281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:18531731 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTAAACTTTTAATGCATATATTT[A/T]AAAAATTCACTTTCCATTTTACTAT
Phenotype
MIM: 601387 MIM: 610985
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
TSG101 PubMed Links

Gene Details

Gene
TSG101
Gene Name
tumor susceptibility 101
There are no transcripts associated with this gene.

Gene
UEVLD
Gene Name
UEV and lactate/malate dehyrogenase domains
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040697.2 1438 UTR 3 NP_001035787.1
NM_001261382.1 1438 UTR 3 NP_001248311.1
NM_001261383.1 1438 UTR 3 NP_001248312.1
NM_001261384.1 1438 UTR 3 NP_001248313.1
NM_001261385.1 1438 UTR 3 NP_001248314.1
NM_001261386.1 1438 UTR 3 NP_001248315.1
NM_001297771.1 1438 Intron NP_001284700.1
NM_018314.4 1438 UTR 3 NP_060784.3
XM_006718255.2 1438 UTR 3 XP_006718318.1
XM_011520201.2 1438 UTR 3 XP_011518503.1
XM_017017985.1 1438 UTR 3 XP_016873474.1
XM_017017986.1 1438 UTR 3 XP_016873475.1

View Full Product Details