Product Details

SNP ID
rs10733026
Assay Type
Functionally tested
NCBI dbSNP Submissions
54
Location
Chr.1:19890130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAGCTGAGGCCCAGGGAAGTTT[A/G]ATGACTCTCCTGAGTCAGCCAGCTG
Phenotype
MIM: 611758
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OTUD3 PubMed Links

Gene Details

Gene
OTUD3
Gene Name
OTU deubiquitinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015207.1 Intron NP_056022.1
XM_005245792.3 Intron XP_005245849.1
XM_005245793.3 Intron XP_005245850.1
XM_005245794.3 Intron XP_005245851.1

View Full Product Details