Product Details

SNP ID
rs11833259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7793515 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAACTCCACTTACCAGGGTAGGAGA[A/G]ACCCTAACTCACACTGGTTCTCATT
Phenotype
MIM: 607937
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NANOG PubMed Links

Gene Details

Gene
NANOG
Gene Name
Nanog homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297698.1 Intron NP_001284627.1
NM_024865.3 Intron NP_079141.2
XM_011520850.1 Intron XP_011519152.1
XM_011520852.1 Intron XP_011519154.1

View Full Product Details