Product Details
- SNP ID
-
rs12412962
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:119149801 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTCAGGGATATAAAGTAAAATTGC[A/T]TGACTTCCGGAAGGGAAAAATGGCT
- Phenotype
-
MIM: 615564
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SFXN4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113691550] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SFXN4
- Gene Name
- sideroflexin 4
View Full Product Details