Product Details

SNP ID
rs12775857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119214459 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTAAGCATAATTAAAAGGTTAT[A/G]CTTGAGGGATAGACCTGAATCTGGA
Phenotype
MIM: 600870
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRK5 PubMed Links

Gene Details

Gene
GRK5
Gene Name
G protein-coupled receptor kinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005308.2 Intron NP_005299.1
XM_005269707.1 Intron XP_005269764.1
XM_005269708.1 Intron XP_005269765.1

View Full Product Details