Product Details

SNP ID
rs11202282
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:86979107 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAACCACACTCAAAAGAAAAAAT[C/T]CAAGTAACCTTGAAAATCAGTATGT
Phenotype
MIM: 608925
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADIRF PubMed Links

Gene Details

Gene
ADIRF
Gene Name
adipogenesis regulatory factor
There are no transcripts associated with this gene.

Gene
AGAP11
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133447.1 Intron NP_597704.1
Gene
MMRN2
Gene Name
multimerin 2
There are no transcripts associated with this gene.

View Full Product Details