Product Details

SNP ID
rs13008067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:187343434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGTGGTTTCTACTAGCAGATTAA[A/T]ACCAAGAGAAAATTAAAAGTAAGTT
Phenotype
MIM: 114190
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CALCRL PubMed Links

Gene Details

Gene
CALCRL
Gene Name
calcitonin receptor like receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271751.1 4600 UTR 3 NP_001258680.1
NM_005795.5 4600 UTR 3 NP_005786.1
XM_005246231.3 4600 UTR 3 XP_005246288.2
XM_005246232.3 4600 UTR 3 XP_005246289.2
XM_005246234.3 4600 UTR 3 XP_005246291.2
XM_017003151.1 4600 UTR 3 XP_016858640.1

View Full Product Details