Product Details

SNP ID
rs7089740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:119897538 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTACTTGGGAATACAAATACCAC[A/G]TGTTCATTTGTACAATAAGTGCTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEC23IP PubMed Links

Gene Details

Gene
SEC23IP
Gene Name
SEC23 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007190.3 Intron NP_009121.1
XM_005269469.2 Intron XP_005269526.1
XM_005269470.2 Intron XP_005269527.1
XM_017015610.1 Intron XP_016871099.1

View Full Product Details