Product Details

SNP ID
rs12601381
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:76310864 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTGGAACCTGGGAAAGTGTTTG[G/T]GTCCAGCCACAGACCTGACTGCTGT
Phenotype
MIM: 601249
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
PRPSAP1 PubMed Links

Gene Details

Gene
PRPSAP1
Gene Name
phosphoribosyl pyrophosphate synthetase associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002766.2 2282 UTR 3 NP_002757.2
XM_011525034.2 2282 Intron XP_011523336.1
Gene
QRICH2
Gene Name
glutamine rich 2
There are no transcripts associated with this gene.

View Full Product Details