Product Details
- SNP ID
-
hCV30916870
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:231370732 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGCCAAATATGTAAGCAGGAGTA[A/C]CCAAAAATGCTACAAGATTAAATTT
- Phenotype
-
MIM: 606425
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EGLN1
PubMed Links
Gene Details
- Gene
- EGLN1
- Gene Name
- egl-9 family hypoxia inducible factor 1
There are no transcripts associated with this gene.
View Full Product Details