Product Details

SNP ID
rs11799453
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:231167077 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGCTGGGTGCAGATGGCCAGCT[C/T]TGAATTAAACTGGGTAGGAGTTGAT
Phenotype
MIM: 610584
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM67 PubMed Links

Gene Details

Gene
TRIM67
Gene Name
tripartite motif containing 67
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004342.3 Intron NP_001004342.3
NM_001300889.1 Intron NP_001287818.1
XM_011544192.2 Intron XP_011542494.1
XM_017001323.1 Intron XP_016856812.1

View Full Product Details