Product Details
- SNP ID
-
rs6849871
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:151282512 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATCCTTACCATGGAAAATATTTTT[C/G]AACAAAACCAGATCAATCTAATGAT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PRSS48
PubMed Links
Gene Details
- Gene
- PRSS48
- Gene Name
- protease, serine 48
View Full Product Details