Product Details

SNP ID
rs12150787
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:6735468 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGACTGTCCTTGTCTTTCATGACA[C/T]TTTTTGAGAACTGGTTGGTGATTTA
Phenotype
MIM: 610592
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP28 PubMed Links

Gene Details

Gene
ARHGAP28
Gene Name
Rho GTPase activating protein 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010000.2 Intron NP_001010000.1
XM_005258144.1 Intron XP_005258201.2
XM_005258145.1 Intron XP_005258202.2
XM_005258146.2 Intron XP_005258203.1
Gene
LOC101927168
Gene Name
uncharacterized LOC101927168
There are no transcripts associated with this gene.

View Full Product Details