Product Details

SNP ID
hCV30926023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:53426602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATAGGTCAGTGCAGGGGAAGAGA[C/T]TAGGGGAGACGGCTGACCCTTCTCC
Phenotype
MIM: 300256 MIM: 300040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HSD17B10 PubMed Links

Gene Details

Gene
HSD17B10
Gene Name
hydroxysteroid 17-beta dehydrogenase 10
There are no transcripts associated with this gene.

Gene
RIBC1
Gene Name
RIB43A domain with coiled-coils 1
There are no transcripts associated with this gene.

Gene
SMC1A
Gene Name
structural maintenance of chromosomes 1A
There are no transcripts associated with this gene.

View Full Product Details