Product Details

SNP ID
rs12337015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:130065901 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCAGCTTTATGCAGTGGATCTTTC[A/C]TAATCCATTGGCTGCAGCCTCACTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GPR107 PubMed Links

Gene Details

Gene
GPR107
Gene Name
G protein-coupled receptor 107
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136557.1 Intron NP_001130029.1
NM_001136558.1 Intron NP_001130030.1
NM_001287346.1 Intron NP_001274275.1
NM_020960.4 Intron NP_066011.2
XM_017014953.1 Intron XP_016870442.1
XM_017014954.1 Intron XP_016870443.1

View Full Product Details