Product Details
- SNP ID
-
rs13139151
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:250167 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGAGTTTTTTTTTGCTTTTTACAA[A/C]CAGTAGTATTTCATTATTTTAATAT
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF876P
PubMed Links
Gene Details
- Gene
- ZNF876P
- Gene Name
- zinc finger protein 876, pseudogene
There are no transcripts associated with this gene.
View Full Product Details