Product Details

SNP ID
rs7695945
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:227834 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAAGTGAAGTAGTACACAGTCTTA[A/C]TTATGTTTTGGAATTCCCACAAAGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC107986244 PubMed Links

Gene Details

Gene
LOC107986244
Gene Name
zinc finger protein 141-like
There are no transcripts associated with this gene.

Gene
ZNF876P
Gene Name
zinc finger protein 876, pseudogene
There are no transcripts associated with this gene.

View Full Product Details