Product Details

SNP ID
rs11147283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:133182543 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTAATGAGTGCTCTGGGAGCAGG[C/T]TGTGGAAATACACGGAGACATGCAG
Phenotype
MIM: 604753
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF268 PubMed Links

Gene Details

Gene
ZNF268
Gene Name
zinc finger protein 268
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165881.2 Intron NP_001159353.1
NM_001165882.2 Intron NP_001159354.1
NM_001165883.1 Intron NP_001159355.1
NM_001165884.2 Intron NP_001159356.2
NM_001165885.1 Intron NP_001159357.1
NM_001165886.1 Intron NP_001159358.1
NM_001165887.1 Intron NP_001159359.1
NM_003415.2 Intron NP_003406.1
NM_152943.2 Intron NP_694422.2

View Full Product Details