Product Details
- SNP ID
-
hCV30939450
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:133132143 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGAAAATAATTTTAGCTAGAAATA[C/T]ATGTGAACTATATGAGAAAAACTTC
- Phenotype
-
MIM: 194538
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF10
PubMed Links
Gene Details
- Gene
- ZNF10
- Gene Name
- zinc finger protein 10
There are no transcripts associated with this gene.
- Gene
- ZNF891
- Gene Name
- zinc finger protein 891
There are no transcripts associated with this gene.
View Full Product Details