Product Details
- SNP ID
-
rs12288569
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:5344556 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCTTTTACTGAAGAGAAGAGATTA[G/T]TCCTTTGAGAAGGTTTTAATATTCC
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OR51B5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113612346] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR51B5
- Gene Name
- olfactory receptor family 51 subfamily B member 5
- Gene
- OR51B6
- Gene Name
- olfactory receptor family 51 subfamily B member 6
There are no transcripts associated with this gene.
View Full Product Details