Product Details
- SNP ID
-
rs12026330
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
9
- Location
-
Chr.1:24058628 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTACATAAATAAGTTCAGTCCTAGA[C/T]GGGCTTTTAGGGATCTCTCAGTAGA
- Phenotype
-
MIM: 616832
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MYOM3
PubMed Links
Gene Details
- Gene
- MYOM3
- Gene Name
- myomesin 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152372.3 |
|
Intron |
|
|
NP_689585.3 |
View Full Product Details