Product Details

SNP ID
rs11134975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:176471996 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCCAGCCCCTATCTGTACATTCT[A/C]ATCACCTGTGTTAAGTTCCCTTTCC
Phenotype
MIM: 616935
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAF2 PubMed Links

Gene Details

Gene
FAF2
Gene Name
Fas associated factor family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014613.2 Intron NP_055428.1
XM_011534475.2 Intron XP_011532777.1

View Full Product Details