Product Details

SNP ID
rs13189533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178152158 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCCCAAACCTACTCGTACCTCT[A/C]TAATCCTTTCTTCAATTGTCAGAAA
Phenotype
MIM: 606470
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NHP2 PubMed Links

Gene Details

Gene
NHP2
Gene Name
NHP2 ribonucleoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001034833.1 Intron NP_001030005.1
NM_017838.3 Intron NP_060308.1
Gene
RMND5B
Gene Name
required for meiotic nuclear division 5 homolog B
There are no transcripts associated with this gene.

View Full Product Details